MAT089757799 Matrix Scientific

4-[2-(4-Methoxy-phenyl)-ethyl]-piperidine

  • MDL number MFCD05022476
  • Cas Number : 654662-60-9
  • Molecular Weight : 219.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020667.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020667.html
Availability details for
Quantity
  • Availability
  • Add to Cart