MAT089757834 Matrix Scientific

4-Amino-2-ethyl-2H-pyrazole-3-carboxylic acid amide

  • MDL number MFCD03422545
  • Cas Number : 957513-91-6
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020703.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020703.html
Availability details for
Quantity
  • Availability
  • Add to Cart