MAT089757840 Matrix Scientific

7-Methoxy-2,3-dihydro-benzo[1,4]dioxine-6-carbaldehyde

  • MDL number MFCD08144775
  • Cas Number : 116757-66-5
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020713.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020713.html
Availability details for
Quantity
  • Availability
  • Add to Cart