MAT089757841 Matrix Scientific

3-(7-Methoxy-2,3-dihydro-benzo[1,4]dioxin-6-yl)-propionic acid

  • MDL number MFCD08144776
  • Cas Number : 883545-08-2
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020714.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020714.html
Availability details for
Quantity
  • Availability
  • Add to Cart