MAT089757852 Matrix Scientific

2-(3-Trifluoromethyl-phenoxy)-propionic acid

  • MDL number MFCD03422213
  • Cas Number : 836-35-1
  • Molecular Weight : 234.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-66
Country of Origin USA
Boiling point 140-144�Torr
Product Link http://www.matrixscientific.com/020726.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020726.html
Availability details for
Quantity
  • Availability
  • Add to Cart