MAT089757853 Matrix Scientific

1,3-Dimethyl-5-methylaminomethyl-1,3-dihydro-benzoimidazol-2-one

  • MDL number MFCD07434398
  • Cas Number : 881450-62-0
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020727.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020727.html
Availability details for
Quantity
  • Availability
  • Add to Cart