MAT089757858 Matrix Scientific

(3,7-Dimethyl-5-oxo-5,8-dihydro-[1,2,4]triazolo-[4,3-a]pyrimidin-6-yl)-acetic acid

  • MDL number MFCD08144846
  • Cas Number : 883550-10-5
  • Molecular Weight : 222.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient
Country of Origin USA
Product Link http://www.matrixscientific.com/020733.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020733.html
Availability details for
Quantity
  • Availability
  • Add to Cart