MAT089757891 Matrix Scientific

(2-Amino-4-oxo-5,6-dihydro-4H-[1,3]thiazin-5-yl)-acetic acid

  • MDL number MFCD00476172
  • Cas Number : 137017-77-7
  • Molecular Weight : 188.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020769.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020769.html
Availability details for
Quantity
  • Availability
  • Add to Cart