MAT089757894 Matrix Scientific

3-(4-Fluoro-phenoxymethyl)-4-methoxy-benzaldehyde

  • MDL number MFCD01114921
  • Cas Number : 329222-88-0
  • Molecular Weight : 260.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020774.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020774.html
Availability details for
Quantity
  • Availability
  • Add to Cart