MAT089757909 Matrix Scientific

3-[3-(3-Methoxy-phenyl)-[1,2,4]oxadiazol-5-yl]-propionic acid

  • MDL number MFCD08133365
  • Cas Number : 500026-33-5
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020789.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020789.html
Availability details for
Quantity
  • Availability
  • Add to Cart