MAT089757937 Matrix Scientific

2-Methyl-2-(4-methyl-piperazin-1-yl)-propylamine

  • MDL number MFCD05201608
  • Cas Number : 21404-92-2
  • Molecular Weight : 171.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020821.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020821.html
Availability details for
Quantity
  • Availability
  • Add to Cart