MAT089757941 Matrix Scientific

2-(4-Methyl-piperazin-1-ylmethyl)-phenylamine

  • MDL number MFCD06804544
  • Cas Number : 19577-84-5
  • Molecular Weight : 205.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86
Country of Origin USA
Product Link http://www.matrixscientific.com/020825.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020825.html
Availability details for
Quantity
  • Availability
  • Add to Cart