MAT089757979 Matrix Scientific

5-Propyl-furan-2-carbaldehyde

  • MDL number MFCD08144844
  • Cas Number : 14497-27-9
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100-101
Density 1.0326/20
Country of Origin USA
Boiling point 100-101� Torr
Product Link http://www.matrixscientific.com/020867.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020867.html
Availability details for
Quantity
  • Availability
  • Add to Cart