MAT089757983 Matrix Scientific

1-(2-Hydroxy-3-pyrrolidin-1-yl-propyl)-2-methyl-1H-indole-3-carbaldehyde

  • MDL number MFCD08144882
  • Cas Number : 883543-75-7
  • Molecular Weight : 286.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020871.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020871.html
Availability details for
Quantity
  • Availability
  • Add to Cart