MAT089757987 Matrix Scientific

2-Piperidino-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD01568826
  • Cas Number : 129865-52-7
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020875.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020875.html
Availability details for
Quantity
  • Availability
  • Add to Cart