MAT089757998 Matrix Scientific

(2,4-Diethoxy-6-methyl-pyrimidin-5-yl)-acetic acid

  • MDL number MFCD07186333
  • Cas Number : 500159-50-2
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020887.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020887.html
Availability details for
Quantity
  • Availability
  • Add to Cart