MAT089758000 Matrix Scientific

2-(2-Chloro-benzyloxy)-ethylamine

  • MDL number MFCD04225849
  • Cas Number : 6594-66-7
  • Molecular Weight : 185.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Boiling point 98-100�1 Torr
Product Link http://www.matrixscientific.com/020889.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020889.html
Availability details for
Quantity
  • Availability
  • Add to Cart