MAT089758028 Matrix Scientific

2-(2-Pyrrolidin-1-yl-ethoxy)-benzaldehyde

  • MDL number MFCD05832990
  • Cas Number : 35166-80-4
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020918.html

Safety Information

MSDS http://www.matrixscientific.com/020918.html
pictograms Irritant/Refrigerate
Availability details for
Quantity
  • Availability
  • Add to Cart