MAT089758037 Matrix Scientific

3-Methoxy-2-[2-(4-methyl-piperazin-1-yl)-ethoxy]-benzaldehyde

  • MDL number MFCD08146604
  • Cas Number : 883545-96-8
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020927.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020927.html
Availability details for
Quantity
  • Availability
  • Add to Cart