MAT089758038 Matrix Scientific

4-[2-(4-Methyl-piperazin-1-yl)-ethoxy]-benzoic acid

  • MDL number MFCD08146605
  • Cas Number : 401804-56-6
  • Molecular Weight : 264.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020928.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020928.html
Availability details for
Quantity
  • Availability
  • Add to Cart