MAT089758039 Matrix Scientific

4-[2-(2-Methyl-piperidin-1-yl)-ethoxy]-benzaldehyde

  • MDL number MFCD08146606
  • Cas Number : 883543-05-3
  • Molecular Weight : 247.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020929.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020929.html
Availability details for
Quantity
  • Availability
  • Add to Cart