MAT089758049 Matrix Scientific

(2-Methyl-3-propionyl-indol-1-yl)-acetic acid

  • MDL number MFCD08146615
  • Cas Number : 883543-84-8
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020939.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020939.html
Availability details for
Quantity
  • Availability
  • Add to Cart