MAT089758057 Matrix Scientific

4-Amino-furazan-3-carboxylic acid methylamide

  • MDL number MFCD08059796
  • Cas Number : 30720-84-4
  • Molecular Weight : 142.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020948.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020948.html
Availability details for
Quantity
  • Availability
  • Add to Cart