MAT089758076 Matrix Scientific

3-(4-Methyl-2-oxo-2H-quinolin-1-yl)-propionic acid

  • MDL number MFCD00436396
  • Cas Number : 75371-21-0
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245-247
Country of Origin USA
Product Link http://www.matrixscientific.com/020405.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020405.html
Availability details for
Quantity
  • Availability
  • Add to Cart