MAT091970526 Matrix Scientific

2-Chloro-5-methylphenyl4-piperidinylmethyl ether hydrochloride

  • MDL number MFCD08445713
  • Cas Number : 1185294-10-3
  • Molecular Weight : 276.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/013467.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/013467.html
Availability details for
Quantity
  • Availability
  • Add to Cart