MAT091970530 Matrix Scientific

4-[(3-Methylphenoxy)methyl]piperidinehydrochloride

  • MDL number MFCD08445785
  • Cas Number : 614731-24-7
  • Molecular Weight : 241.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/013717.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/013717.html
Availability details for
Quantity
  • Availability
  • Add to Cart