MAT091970532 Matrix Scientific

4-[(2,3-Dihydro-1H-inden-5-yloxy)methyl]piperidinehydrochloride

  • MDL number MFCD08445728
  • Cas Number : 1185294-43-2
  • Molecular Weight : 267.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/013966.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/013966.html
Availability details for
Quantity
  • Availability
  • Add to Cart