MAT091970539 Matrix Scientific

(2-Chloro-4-methoxyphenyl)acetonitrile

  • MDL number MFCD08445710
  • Cas Number : 170737-93-6
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-44
Purity 95
Country of Origin USA
Product Link http://www.matrixscientific.com/021049.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021049.html
Availability details for
Quantity
  • Availability
  • Add to Cart