MAT091970544 Matrix Scientific

4-Bromo-2,3-difluorophenol

  • MDL number MFCD08061907
  • Cas Number : 144292-32-0
  • Molecular Weight : 209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/021128.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021128.html
Availability details for
Quantity
  • Availability
  • Add to Cart