MAT091970560 Matrix Scientific

2-([1,1'-Biphenyl]-4-yloxy)ethylaminehydrochloride

  • MDL number MFCD08445694
  • Cas Number : 1051368-98-9
  • Molecular Weight : 249.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/013801.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/013801.html
Availability details for
Quantity
  • Availability
  • Add to Cart