MAT091970578 Matrix Scientific

4-Pyrrolidin-1-yl-pentanoic acid

  • MDL number MFCD08445807
  • Cas Number : 889940-05-0
  • Molecular Weight : 171.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021078.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021078.html
Availability details for
Quantity
  • Availability
  • Add to Cart