MAT091970581 Matrix Scientific

1-(3,4,5-Trimethoxy-phenyl)-ethylamine

  • MDL number MFCD02656660
  • Cas Number : 121082-99-3
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021082.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021082.html
Availability details for
Quantity
  • Availability
  • Add to Cart