MAT091970615 Matrix Scientific

4-Fluoro-3-methylbenzonitrile

  • MDL number MFCD03788510
  • Cas Number : 185147-08-4
  • Molecular Weight : 135.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.matrixscientific.com/021120.html

Safety Information

hcodes Toxic
MSDS http://www.matrixscientific.com/021120.html
Availability details for
Quantity
  • Availability
  • Add to Cart