MAT091970623 Matrix Scientific

2-(Allyloxy)aniline hydrochloride

  • MDL number MFCD08134362
  • Cas Number : 366020-27-1
  • Molecular Weight : 185.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/017100.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/017100.html
Availability details for
Quantity
  • Availability
  • Add to Cart