MAT091970629 Matrix Scientific

alpha,alpha-Difluorophenylacetic acid ethyl ester

  • MDL number MFCD08275296
  • Cas Number : 2248-46-6
  • Molecular Weight : 200.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin USA
Boiling point 70-72�m
Product Link http://www.matrixscientific.com/021136.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021136.html
Availability details for
Quantity
  • Availability
  • Add to Cart