MAT091970631 Matrix Scientific

2-(4-Fluorophenoxy)-N-methyl-1-ethanaminehydrochloride

  • MDL number MFCD08445756
  • Cas Number : 1051919-28-8
  • Molecular Weight : 205.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021138.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021138.html
Availability details for
Quantity
  • Availability
  • Add to Cart