MAT100860033 Matrix Scientific

2-(3,5-Dimethyl-pyrazol-1-yl)-propylaminedihydrochloride

  • MDL number MFCD08687687
  • Cas Number : 1002111-01-4
  • Molecular Weight : 226.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020816.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020816.html
Availability details for
Quantity
  • Availability
  • Add to Cart