MAT100860034 Matrix Scientific

1-Phenethyl-1H-benzoimidazol-5-ylaminedihydrochloride

  • MDL number MFCD08688457
  • Cas Number : 1179363-13-3
  • Molecular Weight : 310.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020890.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020890.html
Availability details for
Quantity
  • Availability
  • Add to Cart