MAT100860037 Matrix Scientific

3-(5-Amino-2-isopropyl-benzoimidazol-1-yl)-propan-1-ol dihydrochloride

  • MDL number MFCD08686807
  • Cas Number : 1185304-23-7
  • Molecular Weight : 306.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020893.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020893.html
Availability details for
Quantity
  • Availability
  • Add to Cart