MAT100860043 Matrix Scientific

[1,4']Bipiperidinyl-3-carboxylic acid ethyl esterdihydrochloride

  • MDL number MFCD08448123
  • Cas Number : 340962-71-2
  • Molecular Weight : 313.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021080.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021080.html
Availability details for
Quantity
  • Availability
  • Add to Cart