MAT100860047 Matrix Scientific

(2-Methoxy-propyl)-methyl-aminehydrochloride

  • MDL number MFCD08448110
  • Cas Number : 883543-35-9
  • Molecular Weight : 139.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021094.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021094.html
Availability details for
Quantity
  • Availability
  • Add to Cart