MAT100860062 Matrix Scientific

1-(3-Methyl-pyridin-2-ylmethyl)-[1,4]diazepanedihydrochloride

  • MDL number MFCD08448124
  • Cas Number : 1185303-21-2
  • Molecular Weight : 278.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021209.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021209.html
Availability details for
Quantity
  • Availability
  • Add to Cart