MAT100860076 Matrix Scientific

2,2-Difluoro-N-(2-hydroxyethyl)propionamide

  • MDL number MFCD08687553
  • Cas Number : 851728-91-1
  • Molecular Weight : 153.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Boiling point 85�2mm
Product Link http://www.matrixscientific.com/021389.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021389.html
Availability details for
Quantity
  • Availability
  • Add to Cart