MAT100860093 Matrix Scientific

7,8-Dimethyl-1,2,3,9b-tetraaza-cyclopenta[a]-naphthalene-4-carbaldehyde

  • MDL number MFCD08445742
  • Cas Number : 889939-65-5
  • Molecular Weight : 226.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021157.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021157.html
Availability details for
Quantity
  • Availability
  • Add to Cart