MAT100860096 Matrix Scientific

7-Chloro-2-(4-chloro-phenyl)-5-phenyl-pyrazolo[1,5-a]pyrimidine

  • MDL number MFCD08445703
  • Cas Number : 889939-44-0
  • Molecular Weight : 340.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021160.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021160.html
Availability details for
Quantity
  • Availability
  • Add to Cart