MAT100860102 Matrix Scientific

1-Phenethyl-1H-[1,2,3]triazole-4-carboxylic acid

  • MDL number MFCD08445800
  • Cas Number : 889940-03-8
  • Molecular Weight : 217.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021166.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021166.html
Availability details for
Quantity
  • Availability
  • Add to Cart