MAT100860106 Matrix Scientific

4-(3-Fluoro-4-methoxy-phenyl)-thiazol-2-ylaminehydrobromide

  • MDL number MFCD06653170
  • Cas Number : 1049742-89-3
  • Molecular Weight : 305.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020832.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020832.html
Availability details for
Quantity
  • Availability
  • Add to Cart