MAT100860113 Matrix Scientific

2-Amino-N-tert-butyl-acetamidehydrochloride

  • MDL number MFCD07324741
  • Cas Number : 71034-40-7
  • Molecular Weight : 166.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/020878.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/020878.html
Availability details for
Quantity
  • Availability
  • Add to Cart