MAT100860120 Matrix Scientific

2-(6-Methyl-pyridin-2-yl)-1-phenyl-ethanonehydrochloride

  • MDL number MFCD00035340
  • Cas Number : 200417-36-3
  • Molecular Weight : 247.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/019934.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/019934.html
Availability details for
Quantity
  • Availability
  • Add to Cart