MAT100860139 Matrix Scientific

C-(2,5-Dimethyl-phenyl)-C-pyridin-4-yl-methylamine

  • MDL number MFCD08445740
  • Cas Number : 889939-63-3
  • Molecular Weight : 212.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://www.matrixscientific.com/021204.html

Safety Information

hcodes Irritant
MSDS http://www.matrixscientific.com/021204.html
Availability details for
Quantity
  • Availability
  • Add to Cart